accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOHOOH

CAS RN 2029-49-4

IUPAC Name: 3-hydroxy-2,6-bis(hydroxymethyl)-4H-pyran-4-one
Synonyms: 3-hydroxy-2,6-bis(hydroxymethyl)pyran-4-one
Molecular Weight (g/mol): 172.14
Molecular Formula: C7H8O5
InChi Key: YZUXYQQFVNGYCA-UHFFFAOYSA-N
SMILES: OCC1=CC(=O)C(O)=C(CO)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results