CAS RN 20290-09-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ Morphine-3-β-D-glucuronide Solution, 1 mg/mL in methanol with 0.05% NaOH, Sold by MilliporeSigma™ Supelco™
A certified solution standard of Morphine-3-β-D-glucuronide at a concentration of 1.0 mg/mL for high volumes of sample testing. This certified solution standard is suitable for LCMS or GCMS applications including forensic analysis or urine drug testing.