accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20291-61-6

IUPAC Name: 1-(methylsulfanyl)heptane
Synonyms: heptanethiol, methyl-
Molecular Weight (g/mol): 146.29
Molecular Formula: C8H18S
InChi Key: FJDWJOQOEZRIDJ-UHFFFAOYSA-N
SMILES: CCCCCCCSC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results