accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 202925-08-4

IUPAC Name: 1-chloro-3-fluoro-5-methoxybenzene
Synonyms: 1-chloro-3-fluoro-5-methoxybenzene
Molecular Weight (g/mol): 160.57
Molecular Formula: C7H6ClFO
InChi Key: XPZBNEWAZPZUHF-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC(Cl)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results