accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNH2

CAS RN 20295-34-5

IUPAC Name: cyclopropanecarbothioamide
Synonyms: cyclopropanecarbothioamide
Molecular Weight (g/mol): 101.17
Molecular Formula: C4H7NS
InChi Key: IIPJWNFOLPDTEQ-UHFFFAOYSA-N
SMILES: NC(=S)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results