accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClONCl

CAS RN 20295-64-1

IUPAC Name: hydrogen 2-chloropyridin-1-ium-1-olate chloride
Synonyms: hydrogen 2-chloropyridin-1-ium-1-olate chloride
Molecular Weight (g/mol): 166
Molecular Formula: C5H5Cl2NO
InChi Key: GRZNODNSNCXOHE-UHFFFAOYSA-N
SMILES: [H+].[Cl-].[O-][N+]1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results