accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFCl

CAS RN 202982-70-5

IUPAC Name: 3-chloro-5-fluorophenol
Synonyms: 3-chloro-5-fluorophenol
Molecular Weight (g/mol): 146.55
Molecular Formula: C6H4ClFO
InChi Key: ZMFBYTAAMHWQHD-UHFFFAOYSA-N
SMILES: OC1=CC(F)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results