accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 202982-77-2

IUPAC Name: 1,2-dibromo-4-fluoro-5-methylbenzene
Synonyms: 1,2-dibromo-4-fluoro-5-methylbenzene
Molecular Weight (g/mol): 267.92
Molecular Formula: C7H5Br2F
InChi Key: GJHJZTCWLKJMIG-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(Br)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results