accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 203-64-5

IUPAC Name: tetracyclo[10.2.1.0⁵,¹⁴.0⁸,¹³]pentadeca-1(14),2,4,6,8,10,12-heptaene
Synonyms: 4,5-methylenephenanthrene
Molecular Weight (g/mol): 190.25
Molecular Formula: C15H10
InChi Key: RKZDZWJDQTZDLD-UHFFFAOYSA-N
SMILES: C1C2=C3C4=C1C=CC=C4C=CC3=CC=C2
Molecular Weight (g/mol) 
  • (1)
  • (1)
Boiling Point 
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results