accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20312-36-1

IUPAC Name: (2S)-2-hydroxy-3-phenylpropanoate
Synonyms: (S)-3-phenyllactate
Molecular Weight (g/mol): 165.17
Molecular Formula: C9H9O3
InChi Key: VOXXWSYKYCBWHO-QMMMGPOBSA-M
SMILES: O[C@@H](CC1=CC=CC=C1)C([O-])=O
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)