CAS RN 2032-65-7
IUPAC Name:
3,5-dimethyl-4-(methylsulfanyl)phenyl N-methylcarbamate
Synonyms:
grandslam mesurol methiocarb certan caswell no. 578B lizetan epa pesticide chemical code 100501 mesurol phenol draza mercaptodimethur
Molecular Weight (g/mol):
225.31
Molecular Formula:
C11H15NO2S
InChi Key:
YFBPRJGDJKVWAH-UHFFFAOYSA-N
SMILES:
CNC(=O)OC1=CC(C)=C(SC)C(C)=C1
1
–
1
of
1
results
1
Methiocarb, SPEX CertiPrep™
CAS: 2032-65-7 Molecular Formula: C11H15NO2S Molecular Weight (g/mol): 225.306 InChI Key: YFBPRJGDJKVWAH-UHFFFAOYSA-N PubChem CID: 16248 ChEBI: CHEBI:38508 IUPAC Name: (3,5-dimethyl-4-methylsulfanylphenyl) N-methylcarbamate SMILES: CC1=CC(=CC(=C1SC)C)OC(=O)NC