accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20328-15-8

IUPAC Name: ethyl 3-methyl-1,2-oxazole-4-carboxylate
Synonyms: ethyl 3-methyl-1,2-oxazole-4-carboxylate
Molecular Weight (g/mol): 155.15
Molecular Formula: C7H9NO3
InChi Key: AEWHICNMVQOWJE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CON=C1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results