accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNOHNCH3

CAS RN 2033-30-9

IUPAC Name: 5,6-dimethyl-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 5,6-dimethyl-1,3-dihydro-1,3-benzodiazol-2-one
Molecular Weight (g/mol): 162.19
Molecular Formula: C9H10N2O
InChi Key: ORWJLFLEIZBRBR-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C2NC(=O)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results