accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 203302-95-8

IUPAC Name: 4-bromo-3,5-difluoroaniline
Synonyms: 4-bromo-3,5-difluoroaniline
Molecular Weight (g/mol): 208.01
Molecular Formula: C6H4BrF2N
InChi Key: DKKUSFDAHRASGO-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(Br)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results