accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFH3CN

CAS RN 203389-28-0

IUPAC Name: 1-butylpyridin-1-ium; tetrafluoroboranuide
Synonyms: 1-butylpyridin-1-ium tetrafluoroborate
Molecular Weight (g/mol): 223.02
Molecular Formula: C9H14BF4N
InChi Key: XLWDQAHXRCBPEE-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCCC[N+]1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results