accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOHSOOOSOOO

CAS RN 20349-39-7

IUPAC Name: disodium 4-hydroxynaphthalene-2,7-disulfonate
Synonyms: disodium 4-hydroxynaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 348.25
Molecular Formula: C10H6Na2O7S2
InChi Key: RJMDNLVCFLKDBY-UHFFFAOYSA-L
SMILES: [Na+].[Na+].OC1=C2C=CC(=CC2=CC(=C1)S([O-])(=O)=O)S([O-])(=O)=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)