CAS RN 2035-89-4
IUPAC Name:
1,1-dimethyl-2-methylidenehydrazine
Synonyms:
formaldehyde, dimethylhydrazone
Molecular Weight (g/mol):
72.11
Molecular Formula:
C3H8N2
InChi Key:
NDNVSJIXYFNRDR-UHFFFAOYSA-N
SMILES:
CN(C)N=C
1
–
1
of
1
results
1
Formaldehyde Dimethylhydrazone 98.0+%, TCI America™
CAS: 2035-89-4 Molecular Formula: C3H8N2 Molecular Weight (g/mol): 72.111 MDL Number: MFCD01753011 InChI Key: NDNVSJIXYFNRDR-UHFFFAOYSA-N PubChem CID: 74857 IUPAC Name: N-methyl-N-(methylideneamino)methanamine SMILES: CN(C)N=C