accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20353-93-9

IUPAC Name: ({[(dimethylamino)methylidene]amino}methylidene)dimethylazanium chloride
Synonyms: ({[(dimethylamino)methylidene]amino}methylidene)dimethylazanium chloride
Molecular Weight (g/mol): 163.65
Molecular Formula: C6H14ClN3
InChi Key: DEIBXAPEZDJDRC-UHFFFAOYSA-M
SMILES: [Cl-].CN(C)C=NC=[N+](C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)