accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20356-45-0

IUPAC Name: 3-(1H-indol-3-yl)-3-oxopropanenitrile
Synonyms: 3-(1H-indol-3-yl)-3-oxopropanenitrile
Molecular Weight (g/mol): 184.2
Molecular Formula: C11H8N2O
InChi Key: KSKBLDDGNWKWKN-UHFFFAOYSA-N
SMILES: O=C(CC#N)C1=CNC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results