CAS RN 2037-81-2
IUPAC Name:
ethyl 5-[(phenoxycarbonyl)amino]-1,2,3-thiadiazole-4-carboxylate
Synonyms:
ethyl 5-[(phenoxycarbonyl)amino]-1,2,3-thiadiazole-4-carboxylate
Molecular Weight (g/mol):
293.3
Molecular Formula:
C12H11N3O4S
InChi Key:
FCPHNPRVMWSGSK-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(NC(=O)OC2=CC=CC=C2)SN=N1
1
–
1
of
1
results
1
Promotions Available
Ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, 98+%
Ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, 98+%
CAS: 2037-81-2 Molecular Formula: C12H11N3O4S Molecular Weight (g/mol): 293.30 MDL Number: MFCD00100230 InChI Key: FCPHNPRVMWSGSK-UHFFFAOYSA-N Synonym: ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate,ethyl 5-phenoxycarbonyl amino-1,2,3-thiadiazole-4-carboxylate,5-n-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester,tos-bb-0370,ethyl 5-phenoxycarbonylamino thiadiazole-4-carboxylate,ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, 98+% PubChem CID: 6917126 IUPAC Name: ethyl 5-(phenoxycarbonylamino)thiadiazole-4-carboxylate SMILES: CCOC(=O)C1=C(NC(=O)OC2=CC=CC=C2)SN=N1