accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 203854-51-7

IUPAC Name: 4-(tert-butoxy)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: 4-(tert-butoxy)-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 397.47
Molecular Formula: C23H27NO5
InChi Key: SKYFRYZPXHNPOK-UHFFFAOYNA-N
SMILES: CC(C)(C)OCC(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results