accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOBr

CAS RN 2039-50-1

IUPAC Name: 5-(bromomethyl)-3-phenyl-1,2-oxazole
Synonyms: 5-(bromomethyl)-3-phenyl-1,2-oxazole
Molecular Weight (g/mol): 238.08
Molecular Formula: C10H8BrNO
InChi Key: ANRMBFFXQKJEIS-UHFFFAOYSA-N
SMILES: BrCC1=CC(=NO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results