accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2039-87-4

IUPAC Name: 1-chloro-2-ethenylbenzene
Synonyms: O-chlorostyrene
Molecular Weight (g/mol): 138.59
Molecular Formula: C8H7Cl
InChi Key: ISRGONDNXBCDBM-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)