accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H2C

CAS RN 2039-89-6

IUPAC Name: 2-ethenyl-1,4-dimethylbenzene
Synonyms: styrene, 2,5-dimethyl-
Molecular Weight (g/mol): 132.21
Molecular Formula: C10H12
InChi Key: DBWWINQJTZYDFK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(C=C)=C1
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)