accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)SH(S)OH(R)OH(R)OHO

CAS RN 20408-97-3

IUPAC Name: (2R,3R,4S,5R)-2,3,4,6-tetrahydroxy-5-sulfanylhexanal
Synonyms: (2R,3R,4S,5R)-2,3,4,6-tetrahydroxy-5-sulfanylhexanal
Molecular Weight (g/mol): 196.22
Molecular Formula: C6H12O5S
InChi Key: IJJLRUSZMLMXCN-SLPGGIOYSA-N
SMILES: OC[C@@H](S)[C@@H](O)[C@H](O)[C@@H](O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results