accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3SOOHOSOOOHSOOH3CCH3CH3SOOOHCH3H3CCH3CH3CH3CH3OH

CAS RN 204190-49-8

IUPAC Name: 5,11,17,23-tetra-tert-butyl-25,26,27,28-tetrahydroxy-2λ⁶,8λ⁶,14λ⁶,20λ⁶-tetrathiapentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaen-2,2,8,8,14,14,20,20-octone
Synonyms: 5,11,17,23-tetra-tert-butyl-25,26,27,28-tetrahydroxy-2λ⁶,8λ⁶,14λ⁶,20λ⁶-tetrathiapentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaen-2,2,8,8,14,14,20,20-octone
Molecular Weight (g/mol): 849.05
Molecular Formula: C40H48O12S4
InChi Key: BOLKLFFOYGXSKT-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=C(O)C(=C1)S(=O)(=O)C1=CC(=CC(=C1O)S(=O)(=O)C1=CC(=CC(=C1O)S(=O)(=O)C1=C(O)C(=CC(=C1)C(C)(C)C)S2(=O)=O)C(C)(C)C)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results