CAS RN 204251-33-2
CAS RN 204251-33-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-D-Glu OAll -OH, Advanced ChemTech
CAS: 204251-33-2 Molecular Formula: C23H23NO6 Molecular Weight (g/mol): 409.4