accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 204260-38-8

OOHOONH(R)CH3

CAS RN 204260-38-8

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-methylphenyl)propanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-methylphenyl)propanoic acid
Molecular Weight (g/mol): 401.46
Molecular Formula: C25H23NO4
InChi Key: UXLHLZHGQPDMJQ-HSZRJFAPSA-N
SMILES: CC1=CC=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)