accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20427-58-1

IUPAC Name: zinc hydroxide
Synonyms: zinc hydroxide
Molecular Weight (g/mol): 101.41
Molecular Formula: H4O2Zn
InChi Key: JQBMTMBJMXRRCJ-UHFFFAOYSA-N
SMILES: O.O.[Zn]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results