accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFFFFFFFFF

CAS RN 2043-47-2

IUPAC Name: 3,3,4,4,5,5,6,6,6-nonafluorohexan-1-ol
Synonyms: 3,3,4,4,5,5,6,6,6-nonafluorohexan-1-ol
Molecular Weight (g/mol): 264.09
Molecular Formula: C6H5F9O
InChi Key: JCMNMOBHVPONLD-UHFFFAOYSA-N
SMILES: OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)