accessibility menu, dialog, popup

UserName

Viewing profile as user:
PNCl

CAS RN 20430-33-5

IUPAC Name: [(4-cyanophenyl)methyl]triphenylphosphanium chloride
Synonyms: [(4-cyanophenyl)methyl]triphenylphosphanium chloride
Molecular Weight (g/mol): 413.88
Molecular Formula: C26H21ClNP
InChi Key: GXMFPJIKPZDNCS-UHFFFAOYSA-M
SMILES: [Cl-].N#CC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results