accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOO

CAS RN 204376-55-6

IUPAC Name: (9H-fluoren-9-yl)methyl 4-oxopiperidine-1-carboxylate
Synonyms: 9H-fluoren-9-ylmethyl 4-oxopiperidine-1-carboxylate
Molecular Weight (g/mol): 321.38
Molecular Formula: C20H19NO3
InChi Key: AMWDMRLGMSQZTK-UHFFFAOYSA-N
SMILES: O=C(OCC1C2=CC=CC=C2C2=CC=CC=C12)N1CCC(=O)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results