accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20442-70-0

CH3CH3H3COOCs

CAS RN 20442-70-0

IUPAC Name: caesium(1+) 2,2-dimethylpropanoate
Synonyms: caesium(1+) pivalate
Molecular Weight (g/mol): 234.03
Molecular Formula: C5H9CsO2
InChi Key: LGVUAXNPXVXCCW-UHFFFAOYSA-M
SMILES: [Cs+].CC(C)(C)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results