accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOHOONHN(S)CH3CH3H3CCH3

CAS RN 2044702-38-5

IUPAC Name: (2S)-4-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)butanoic acid
Synonyms: (2S)-4-[(tert-butoxycarbonyl)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}butanoic acid
Molecular Weight (g/mol): 454.52
Molecular Formula: C25H30N2O6
InChi Key: XWVWNUSHPLVJFI-NRFANRHFSA-N
SMILES: CN([C@@H](CCNC(=O)OC(C)(C)C)C(O)=O)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)