accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NCO

CAS RN 20458-99-5

IUPAC Name: 1,3-diethyl-2-isocyanatobenzene
Synonyms: 1,3-diethyl-2-isocyanatobenzene
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: AUSRFKVPUCQCCN-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(CC)=C1N=C=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)