accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 204587-95-1

IUPAC Name: (2R,3S)-4-(tert-butoxy)-3-hydroxy-4-oxo-1-phenylbutan-2-aminium
Synonyms: (2R,3S)-4-(tert-butoxy)-3-hydroxy-4-oxo-1-phenylbutan-2-aminium
Molecular Weight (g/mol): 252.33
Molecular Formula: C14H22NO3
InChi Key: ITPFEPDHSLWUED-NEPJUHHUSA-O
SMILES: CC(C)(C)OC(=O)[C@@H](O)[C@H]([NH3+])CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)