accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20461-99-8

IUPAC Name: ethyl 1,3-dithiolane-2-carboxylate
Synonyms: ethyl 1,3-dithiolane-2-carboxylate
Molecular Weight (g/mol): 178.26
Molecular Formula: C6H10O2S2
InChi Key: OMCSHTHLIQOHDD-UHFFFAOYSA-N
SMILES: CCOC(=O)C1SCCS1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)