accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 204841-19-0

IUPAC Name: (3-acetylphenyl)boronic acid
Synonyms: 3-acetylphenylboronic acid
Molecular Weight (g/mol): 163.97
Molecular Formula: C8H9BO3
InChi Key: SJGGDZCTGBKBCK-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC(=C1)B(O)O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)