accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20487-40-5

H3COOCH3CH3H3C

CAS RN 20487-40-5

IUPAC Name: tert-butyl propanoate
Synonyms: tert-butyl propionate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: JAELLLITIZHOGQ-UHFFFAOYSA-N
SMILES: CCC(=O)OC(C)(C)C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)