accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2OOCH3

CAS RN 2049-80-1

IUPAC Name: 1,3-diethyl 2-(prop-2-en-1-yl)propanedioate
Synonyms: 1,3-diethyl 2-(prop-2-en-1-yl)propanedioate
Molecular Weight (g/mol): 200.23
Molecular Formula: C10H16O4
InChi Key: GDWAYKGILJJNBB-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CC=C)C(=O)OCC
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)