accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20491-53-6

IUPAC Name: bis(propan-2-yl)phosphane
Synonyms: diisopropylphosphane
Molecular Weight (g/mol): 118.16
Molecular Formula: C6H15P
InChi Key: WDIIYWASEVHBBT-UHFFFAOYSA-N
SMILES: CC(C)PC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)