accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 205-82-3

IUPAC Name: benzo[j]fluoranthene
Synonyms: benzo(J)fluoranthene benzo(j)fluoranthene
Molecular Weight (g/mol): 252.32
Molecular Formula: C20H12
InChi Key: KHNYNFUTFKJLDD-UHFFFAOYSA-N
SMILES: C1=CC=C2C3=C(C=CC2=C1)C1=C2C3=CC=CC2=CC=C1
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results