accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHNNO(E)

CAS RN 2050-14-8

IUPAC Name: (6E)-6-[2-(2-hydroxyphenyl)hydrazin-1-ylidene]cyclohexa-2,4-dien-1-one
Synonyms: (6E)-6-[2-(2-hydroxyphenyl)hydrazin-1-ylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 214.22
Molecular Formula: C12H10N2O2
InChi Key: JLRUNUCYUPAYKT-GXDHUFHOSA-N
SMILES: OC1=CC=CC=C1N\N=C1/C=CC=CC1=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)