accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2050-23-9

IUPAC Name: 1,8-diethyl octanedioate
Synonyms: diethyl suberate
Molecular Weight (g/mol): 230.3
Molecular Formula: C12H22O4
InChi Key: PEUGOJXLBSIJQS-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCCC(=O)OCC
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)