accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHCH3H3C

CAS RN 2050-43-3

IUPAC Name: N-(2,4-dimethylphenyl)acetamide
Synonyms: 2,4-dimethylacetanilide
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: PJEIIBXJUDOMAN-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C)C=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results