CAS RN 2050-60-4
IUPAC Name:
dibutyl oxalate
Synonyms:
dibutyl oxalate
Molecular Weight (g/mol):
202.25
Molecular Formula:
C10H18O4
InChi Key:
JKRZOJADNVOXPM-UHFFFAOYSA-N
SMILES:
CCCCOC(=O)C(=O)OCCCC
1
–
1
of
1
results
1
Dibutyl Oxalate 99.0+%, TCI America™
CAS: 2050-60-4 Molecular Formula: C10H18O4 Molecular Weight (g/mol): 202.25 MDL Number: MFCD00009442 InChI Key: JKRZOJADNVOXPM-UHFFFAOYSA-N Synonym: Oxalic Acid Dibutyl Ester PubChem CID: 16306 IUPAC Name: dibutyl oxalate SMILES: CCCCOC(=O)C(=O)OCCCC