accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2050-68-2

IUPAC Name: 4,4'-dichloro-1,1'-biphenyl
Synonyms: polychlorobiphenyl p,p-dcbp p,p-DCBP 4-dcb cpd
Molecular Weight (g/mol): 223.1
Molecular Formula: C12H8Cl2
InChi Key: YTBRNEUEFCNVHC-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=CC=C(Cl)C=C1
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results