accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3SSCH3CH3

CAS RN 2051-04-9

IUPAC Name: 3-methyl-1-[(3-methylbutyl)disulfanyl]butane
Synonyms: disulfide, bis(3-methylbutyl)
Molecular Weight (g/mol): 206.41
Molecular Formula: C10H22S2
InChi Key: MPYGLNNTOXLWOB-UHFFFAOYSA-N
SMILES: CC(C)CCSSCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results