accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClClClClClClCl(?)

CAS RN 2051-24-3

IUPAC Name: decachloro-1,1'-biphenyl
Synonyms: decachlorobiphenyl
Molecular Weight (g/mol): 498.63
Molecular Formula: C12Cl10
InChi Key: ONXPZLFXDMAPRO-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C(Cl)=C(C(Cl)=C1Cl)C1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl
Solvent or Matrix 
  • (1)
  • (1)
Technique 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results