accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 2051-97-0

IUPAC Name: 1-benzyl-1H-pyrrole
Synonyms: benzopyrrole PYRROLE1benzyl
Molecular Weight (g/mol): 157.22
Molecular Formula: C11H11N
InChi Key: FNEQHKCQXDKYEO-UHFFFAOYSA-N
SMILES: C(N1C=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)